siRNA Details
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008939.2 | NP_032965.1 | 4 | 1183 | ||
GenBank mRNA | AK132633.1 | - | 4 | 972 | ||
BC031429.1 | - | 4 | 1085 | 3665834 | ||
D89871.1 | - | 4 | 1181 | |||
Y13192.1 | - | 4 | 960 |
