siRNA Details
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008939.2 | NP_032965.1 | 3 | 894 | ||
GenBank mRNA | AK132633.1 | - | 3 | 683 | ||
BC031429.1 | - | 3 | 796 | 3665834 | ||
D89871.1 | - | 3 | 892 | |||
Y13192.1 | - | 3 | 671 |
