siRNA Details
| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011176.4 | NP_035306.2 | 3, 4 | 549 | ||
| GenBank mRNA | AF042822.3 | - | 3, 4 | 438 | ||
| AK052738.1 | - | 3, 4 | 549 | |||
| AK085410.1 | - | 3, 4 | 548 | |||
| AK166745.1 | - | 3, 4 | 540 | |||
| BC005496.1 | - | 3, 4 | 548 | 3488059 | ||
| BC008514.1 | - | 3, 4 | 548 | |||

St14
Genomic Map