siRNA Details
| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011176.4 | NP_035306.2 | 2 | 401 | ||
| GenBank mRNA | AF042822.3 | - | 2 | 290 | ||
| AK052738.1 | - | 2 | 401 | |||
| AK085410.1 | - | 2 | 399 | |||
| AK166745.1 | - | 2 | 392 | |||
| BC005496.1 | - | 2 | 400 | 3488059 | ||
| BC008514.1 | - | 2 | 400 | |||

St14
Genomic Map