siRNA Details
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008941.3 | NP_032967.1 | 14 | 1574 | ||
NM_178855.4 | NP_849186.2 | 13 | 1529 | |||
GenBank mRNA | AK033582.1 | - | 13 | 1531 | ||
AK038356.1 | - | 5 | 523 | |||
BC117917.1 | - | 14 | 1573 | 40097367 | ||
BC117918.1 | - | 14 | 1573 | 40097370 | ||
U73378.1 | - | 14 | 1569 |
