siRNA Details
| Gene Symbol: Psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008943.2 | NP_032969.1 | 8 | 1342 | ||
| GenBank mRNA | AF149111.1 | - | 6 | 854 | ||
| AK134232.1 | - | 8 | 1025 | |||
| AK134615.1 | - | 8 | 1064 | |||
| AK148101.1 | - | 8 | 1063 | |||
| AK165984.1 | - | 8 | 1063 | |||
| AK169963.1 | - | 4 | 469 | |||
| BC030409.1 | - | 8 | 1017 | 5133302 | ||
| BC071233.1 | - | 8 | 1081 | 3673609 | ||
| L42177.1 | - | 8 | 1038 | |||

Psen1
Genomic Map