siRNA Details
| Gene Symbol: Psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008943.2 | NP_032969.1 | 3 | 517 | ||
| GenBank mRNA | AK134232.1 | - | 3 | 200 | ||
| AK134615.1 | - | 3 | 239 | |||
| AK148101.1 | - | 3 | 238 | |||
| AK165984.1 | - | 3 | 238 | |||
| BC014744.1 | - | 3 | 268 | 4511140 | ||
| BC030409.1 | - | 3 | 192 | 5133302 | ||
| BC071233.1 | - | 3 | 256 | 3673609 | ||
| L42177.1 | - | 3 | 213 | |||

Psen1
Genomic Map