siRNA Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128605.1 | NP_001122077.1 | 4, 5 | 588 | ||
| NM_011183.3 | NP_035313.2 | 4, 5 | 597 | |||
| GenBank mRNA | AF038935.1 | - | 3, 4 | 525 | ||
| AK014706.1 | - | 3, 4 | 532 | |||
| AK148111.1 | - | 4, 5 | 589 | |||
| AK154119.1 | - | 4, 5 | 533 | |||
| BC010403.1 | - | 3, 4 | 543 | 4038761 | ||
| U57324.1 | - | 4, 5 | 548 | |||

Psen2
Genomic Map