siRNA Details
| Gene Symbol: Pstpip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013831.4 | NP_038859.3 | 825 | |||
| GenBank mRNA | AK132418.1 | - | 807 | |||
| AK154284.1 | - | 792 | ||||
| AK154724.1 | - | 10 | 831 | |||
| AK157994.1 | - | 10 | 809 | |||
| BC002123.1 | - | 806 | 3494357 | |||
| BC057654.1 | - | 767 | 4460817 | |||
| CT010308.1 | - | 10 | 657 | |||
| Y18101.1 | - | 10 | 657 | |||

Pstpip2
Genomic Map