siRNA Details
| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 18 | 3009 | ||
| GenBank mRNA | AK147626.1 | - | 19 | 3156 | ||
| U46155.1 | - | 18 | 3009 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptch1| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 18 | 3009 | ||
| GenBank mRNA | AK147626.1 | - | 19 | 3156 | ||
| U46155.1 | - | 18 | 3009 | |||