siRNA Details
| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 16, 17 | 2655 | ||
| GenBank mRNA | AK147626.1 | - | 17, 18 | 2802 | ||
| U46155.1 | - | 16, 17 | 2655 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptch1| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 16, 17 | 2655 | ||
| GenBank mRNA | AK147626.1 | - | 17, 18 | 2802 | ||
| U46155.1 | - | 16, 17 | 2655 | |||