siRNA Details
Gene Symbol: Ptdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008959.3 | NP_032985.2 | 13 | 2052 | ||
GenBank mRNA | AF042731.1 | - | 13 | 1467 | ||
AK036990.1 | - | 13 | 1514 | |||
AK054101.1 | - | 13 | 1491 | |||
AK085857.1 | - | 13 | 2053 | |||
AK086136.1 | - | 13 | 1711 | |||
AK088048.1 | - | 13 | 1534 | |||
AK137681.1 | - | 13 | 1722 | |||
AK172873.1 | - | 12 | 3838 | |||
BC003260.1 | - | 13 | 1689 | 3501141 |
Gene Symbol: 4933433G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017041.1 | - | 9 | 849 |
