siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008960.2 | NP_032986.1 | 8 | 1772 | ||
| GenBank mRNA | AK030750.1 | - | 4 | 535 | ||
| AK076980.1 | - | 8 | 1730 | |||
| AK088717.1 | - | 8 | 1751 | |||
| AK138025.1 | - | 1 | 81 | |||
| AK148736.1 | - | 8 | 1607 | |||
| BC021445.1 | - | 8 | 1121 | 5038272 | ||
| U92437.1 | - | 8 | 1852 | |||

Pten
Genomic Map