siRNA Details
| Gene Symbol: Sugct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138654.3 | NP_619595.3 | 7 | 538 | ||
| GenBank mRNA | AF397014.1 | - | 7 | 554 | ||
| AK160265.1 | - | 6 | 411 | |||
| BC055852.1 | - | 7 | 512 | 4221852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sugct| Gene Symbol: Sugct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138654.3 | NP_619595.3 | 7 | 538 | ||
| GenBank mRNA | AF397014.1 | - | 7 | 554 | ||
| AK160265.1 | - | 6 | 411 | |||
| BC055852.1 | - | 7 | 512 | 4221852 | ||