siRNA Details
| Gene Symbol: Sugct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138654.3 | NP_619595.3 | 10, 11 | 911 | ||
| GenBank mRNA | AA108199.1 | - | 4, 5 | 363 | ||
| AK160265.1 | - | 9, 10 | 784 | |||
| BC055852.1 | - | 10, 11 | 885 | 4221852 | ||

Sugct
Genomic Map