siRNA Details
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138655.1 | NP_619596.1 | 12 | 1845 | ||
GenBank mRNA | AF417581.1 | - | 12 | 1845 | ||
AY263156.1 | - | 1 | 12 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138655.1 | NP_619596.1 | 12 | 1845 | ||
GenBank mRNA | AF417581.1 | - | 12 | 1845 | ||
AY263156.1 | - | 1 | 12 |