siRNA Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138656.2 | NP_619597.2 | 7, 8 | 957 | ||
| GenBank mRNA | AJ309922.1 | - | 7, 8 | 914 | ||
| AK089354.1 | - | 7, 8 | 959 | |||
| AK134803.1 | - | 7, 8 | 946 | |||
| AK153439.1 | - | 8, 9 | 1035 | |||
| AK154883.1 | - | 7, 8 | 959 | |||
| AK170007.1 | - | 7, 8 | 896 | |||
| AK170773.1 | - | 7, 8 | 953 | |||
| BC008526.1 | - | 7, 8 | 925 | 3495424 | ||

Mvd
Genomic Map