siRNA Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138656.2 | NP_619597.2 | 8 | 992 | ||
| GenBank mRNA | AJ309922.1 | - | 8 | 949 | ||
| AK089354.1 | - | 8 | 994 | |||
| AK134803.1 | - | 8 | 981 | |||
| AK153439.1 | - | 9 | 1070 | |||
| AK154883.1 | - | 8 | 994 | |||
| AK170007.1 | - | 8 | 931 | |||
| AK170773.1 | - | 8 | 988 | |||
| BC008526.1 | - | 8 | 960 | 3495424 | ||

Mvd
Genomic Map