siRNA Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138656.2 | NP_619597.2 | 7 | 801 | ||
| GenBank mRNA | AJ309922.1 | - | 7 | 758 | ||
| AK089354.1 | - | 7 | 803 | |||
| AK134803.1 | - | 7 | 790 | |||
| AK153439.1 | - | 8 | 879 | |||
| AK154883.1 | - | 7 | 803 | |||
| AK170007.1 | - | 7 | 740 | |||
| AK170773.1 | - | 7 | 797 | |||
| BC008526.1 | - | 7 | 769 | 3495424 | ||

Mvd
Genomic Map