siRNA Details
| Gene Symbol: Socs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138657.3 | NP_619598.1 | 7 | 1540 | ||
| GenBank mRNA | AF424814.1 | - | 7 | 1541 | ||
| AK147580.1 | - | 7 | 1638 | |||
| AK147650.1 | - | 7 | 1638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs7| Gene Symbol: Socs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138657.3 | NP_619598.1 | 7 | 1540 | ||
| GenBank mRNA | AF424814.1 | - | 7 | 1541 | ||
| AK147580.1 | - | 7 | 1638 | |||
| AK147650.1 | - | 7 | 1638 | |||