siRNA Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138665.2 | NP_619606.1 | 12, 13 | 2234 | ||
| GenBank mRNA | AK087163.1 | - | 5, 6 | 535 | ||
| AK140451.1 | - | 11, 12 | 1510 | |||
| AK159236.1 | - | 12, 13 | 2198 | |||
| AK159681.1 | - | 12, 13 | 2198 | |||
| AK159704.1 | - | 12, 13 | 2198 | |||
| AK163515.1 | - | 12, 13 | 1763 | |||
| AK163660.1 | - | 11, 12 | 1510 | |||
| AK182037.1 | - | 1, 2 | 37 | |||
| AK208205.1 | - | 1, 2 | 37 | |||
| BC003456.1 | - | 12, 13 | 1743 | 2648344 | ||

Sardh
Genomic Map