siRNA Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138665.2 | NP_619606.1 | 16 | 2643 | ||
| GenBank mRNA | AK087163.1 | - | 8 | 830 | ||
| AK140451.1 | - | 15 | 1920 | |||
| AK159236.1 | - | 16 | 2607 | |||
| AK159681.1 | - | 16 | 2607 | |||
| AK159704.1 | - | 16 | 2607 | |||
| AK163515.1 | - | 16 | 2172 | |||
| AK163660.1 | - | 15 | 1919 | |||
| AK176928.1 | - | 4 | 425 | |||
| AK178634.1 | - | 4 | 391 | |||
| AK183193.1 | - | 4 | 379 | |||
| BC003456.1 | - | 16 | 2152 | 2648344 | ||

Sardh
Genomic Map