siRNA Details
| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138666.3 | NP_619607.2 | 4 | 1808 | ||
| GenBank mRNA | AK122433.1 | - | 4 | 1809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlgn1| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138666.3 | NP_619607.2 | 4 | 1808 | ||
| GenBank mRNA | AK122433.1 | - | 4 | 1809 | ||