siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290421.1 | NP_001277350.1 | 20 | 3180 | ||
| NM_010227.3 | NP_034357.2 | 20 | 3182 | |||
| GenBank mRNA | AK089311.1 | - | 20 | 3060 | ||
| AK131179.1 | - | 8 | 1023 | |||
| AK152579.1 | - | 20 | 2838 | |||
| BC038478.1 | - | 20 | 3055 | |||
| BC151024.1 | - | 20 | 3173 | 9087937 | ||

Flna
Genomic Map