siRNA Details
| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138672.2 | NP_619613.2 | 44, 45 | 4798 | ||
| GenBank mRNA | AB052959.1 | - | 44, 45 | 4719 | ||
| AF290914.2 | - | 44, 45 | 4729 | |||
| BC031166.1 | - | 7, 8 | 652 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stab1| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138672.2 | NP_619613.2 | 44, 45 | 4798 | ||
| GenBank mRNA | AB052959.1 | - | 44, 45 | 4719 | ||
| AF290914.2 | - | 44, 45 | 4729 | |||
| BC031166.1 | - | 7, 8 | 652 | |||