siRNA Details
Gene Symbol: Ash1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138679.5 | NP_619620.3 | 2 | 1281 | ||
GenBank mRNA | AB221625.1 | - | 1 | 314 | ||
AK088497.1 | - | 3 | 1306 | |||
AK153783.1 | - | 1 | 460 | |||
BC057142.1 | - | 1 | 615 | |||
BC066813.1 | - | 2 | 855 |
