siRNA Details
| Gene Symbol: Ash1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138679.5 | NP_619620.3 | 2 | 3016 | ||
| GenBank mRNA | AB221625.1 | - | 1 | 2049 | ||
| AK153783.1 | - | 1 | 2195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ash1l| Gene Symbol: Ash1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138679.5 | NP_619620.3 | 2 | 3016 | ||
| GenBank mRNA | AB221625.1 | - | 1 | 2049 | ||
| AK153783.1 | - | 1 | 2195 | |||