siRNA Details
| Gene Symbol: Ptgis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008968.3 | NP_032994.1 | 6 | 894 | ||
| GenBank mRNA | AB001607.1 | - | 6 | 812 | ||
| AK048059.1 | - | 947 | ||||
| AK076585.1 | - | 6 | 895 | |||
| AY285779.1 | - | 2 | 243 | |||
| BC062151.1 | - | 6 | 901 | 6512164 | ||

Ptgis
Genomic Map