siRNA Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008969.4 | NP_032995.1 | 7 | 840 | ||
| GenBank mRNA | AK046457.1 | - | 7 | 767 | ||
| AK134163.1 | - | 7 | 726 | |||
| AK138432.1 | - | 7 | 779 | |||
| AK142741.1 | - | 7 | 767 | |||
| AK159907.1 | - | 7 | 841 | |||
| AK160886.1 | - | 7 | 759 | |||
| AK163609.1 | - | 7 | 767 | |||
| AK167357.1 | - | 6 | 718 | |||
| AK170414.1 | - | 7 | 735 | |||
| AK170664.1 | - | 7 | 767 | |||
| BC005573.1 | - | 7 | 757 | 3968101 | ||
| BC023322.1 | - | 7 | 749 | 3968136 | ||
| CT010314.1 | - | 6 | 703 | |||
| M34141.1 | - | 7 | 739 | |||

Ptgs1
Genomic Map