siRNA Details
| Gene Symbol: Pthlh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008970.3 | NP_032996.2 | 3 | 703 | ||
| GenBank mRNA | AJ278119.1 | - | 2 | 198 | ||
| AY183377.1 | - | 2 | 217 | |||
| AY220497.1 | - | 3 | 464 | |||
| BC058187.1 | - | 3 | 508 | 5346064 | ||

Pthlh
Genomic Map