siRNA Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138758.1 | NP_620097.1 | 5 | 858 | ||
| GenBank mRNA | AK052425.1 | - | 5 | 860 | ||
| AK144627.1 | - | 5 | 854 | |||
| AK161724.1 | - | 5 | 825 | |||
| AY033513.1 | - | 4 | 696 | |||
| BC010495.1 | - | 5 | 858 | 4195680 | ||
| BC115365.1 | - | 5 | 790 | 40059106 | ||

Tmlhe
Genomic Map