siRNA Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138758.1 | NP_620097.1 | 7 | 1158 | ||
| GenBank mRNA | AK144627.1 | - | 7 | 1154 | ||
| AK161724.1 | - | 7 | 1125 | |||
| AY033513.1 | - | 6 | 996 | |||
| BC010495.1 | - | 7 | 1158 | 4195680 | ||
| BC115365.1 | - | 7 | 1090 | 40059106 | ||

Tmlhe
Genomic Map