siRNA Details
| Gene Symbol: Ptpn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011203.2 | NP_035333.2 | 8 | 717 | ||
| GenBank mRNA | AK154107.1 | - | 8 | 809 | ||
| BC051980.1 | - | 8 | 717 | 6402545 | ||
| X63440.1 | - | 8 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn12| Gene Symbol: Ptpn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011203.2 | NP_035333.2 | 8 | 717 | ||
| GenBank mRNA | AK154107.1 | - | 8 | 809 | ||
| BC051980.1 | - | 8 | 717 | 6402545 | ||
| X63440.1 | - | 8 | 718 | |||