siRNA Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 3, 4 | 870 | ||
GenBank mRNA | AF170902.1 | - | 2, 3 | 437 | ||
AF170903.1 | - | 2, 3 | 437 | |||
AK028783.1 | - | 3, 4 | 834 | |||
AK037646.1 | - | 3, 4 | 871 | |||
AK086524.1 | - | 3, 4 | 864 | |||
AK156480.1 | - | 3, 4 | 616 | |||
D31842.1 | - | 2, 3 | 467 |
