siRNA Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008976.2 | NP_033002.2 | 14 | 3128 | ||
| GenBank mRNA | AF170903.1 | - | 13 | 1675 | ||
| AK037646.1 | - | 14 | 3129 | |||
| AK156480.1 | - | 14 | 2874 | |||
| D31842.1 | - | 13 | 2725 | |||

Ptpn14
Genomic Map