siRNA Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127177.1 | NP_001120649.1 | 5 | 536 | ||
| NM_008977.3 | NP_033003.1 | 5 | 536 | |||
| GenBank mRNA | AK014528.1 | - | 5 | 504 | ||
| AK076072.1 | - | 5 | 489 | |||
| AK132013.1 | - | 5 | 499 | |||
| BC008269.1 | - | 5 | 510 | 3602983 | ||
| M80739.1 | - | 5 | 491 | |||
| M81477.1 | - | 5 | 491 | |||
| S52655.1 | - | 5 | 486 | |||

Ptpn2
Genomic Map