siRNA Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 7 | 680 | ||
| GenBank mRNA | AK017003.1 | - | 7 | 743 | ||
| AK029493.1 | - | 7 | 678 | |||
| BC139817.1 | - | 6 | 657 | |||
| D64141.1 | - | 6 | 679 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn20| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 7 | 680 | ||
| GenBank mRNA | AK017003.1 | - | 7 | 743 | ||
| AK029493.1 | - | 7 | 678 | |||
| BC139817.1 | - | 6 | 657 | |||
| D64141.1 | - | 6 | 679 | |||