siRNA Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008980.2 | NP_033006.2 | 8, 9 | 1195 | ||
| GenBank mRNA | M36033.1 | - | 8, 9 | 933 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpra| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008980.2 | NP_033006.2 | 8, 9 | 1195 | ||
| GenBank mRNA | M36033.1 | - | 8, 9 | 933 | ||