siRNA Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163688.1 | NP_001157160.1 | 10, 11 | 1293 | ||
| NM_008980.2 | NP_033006.2 | 11, 12 | 1401 | |||
| GenBank mRNA | AF332081.1 | - | 9, 10 | 1006 | ||
| AF332082.1 | - | 9, 10 | 1006 | |||
| AJ431367.1 | - | 9, 10 | 1006 | |||
| AK031690.1 | - | 10, 11 | 1292 | |||
| AK165673.1 | - | 10, 11 | 1291 | |||
| BC050930.1 | - | 10, 11 | 1275 | 6811172 | ||
| BC110683.1 | - | 10, 11 | 1259 | 3965311 | ||
| M36033.1 | - | 11, 12 | 1139 | |||
| Z23054.1 | - | 1, 2 | 43 | |||

Ptpra
Genomic Map