siRNA Details
| Gene Symbol: Wdr81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138950.2 | NP_620400.2 | 10 | 6018 | ||
| GenBank mRNA | AK131146.1 | - | 10 | 2563 | ||
| AK138745.1 | - | 4 | 1076 | |||
| AK143628.1 | - | 9 | 2107 | |||
| AK160585.1 | - | 9 | 2060 | |||
| AK172019.1 | - | 9 | 2204 | |||
| BC025229.1 | - | 4 | 748 | |||
| BC054822.1 | - | 1 | 613 | |||
| BC061040.1 | - | 3 | 555 | |||
| BC080206.1 | - | 4 | 845 | |||

Wdr81
Genomic Map