siRNA Details
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138952.3 | NP_620402.1 | 3 | 497 | ||
| GenBank mRNA | AF461040.1 | - | 3 | 395 | ||
| AF487539.1 | - | 3 | 395 | |||
| AK159593.1 | - | 3 | 521 | |||
| AK172319.1 | - | 3 | 519 | |||
| AK172417.1 | - | 3 | 519 | |||
| BC050845.1 | - | 3 | 510 | |||
| BC058088.1 | - | 3 | 642 | |||
| BC069878.1 | - | 3 | 522 | 5316066 | ||

Ripk2
Genomic Map