siRNA Details
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138952.3 | NP_620402.1 | 4 | 666 | ||
| GenBank mRNA | AF461040.1 | - | 4 | 564 | ||
| AF487539.1 | - | 4 | 564 | |||
| AK159593.1 | - | 4 | 690 | |||
| AK172319.1 | - | 4 | 688 | |||
| AK172417.1 | - | 4 | 688 | |||
| BC025611.1 | - | 2 | 184 | |||
| BC050845.1 | - | 4 | 679 | |||
| BC058088.1 | - | 4 | 811 | |||
| BC069878.1 | - | 4 | 691 | 5316066 | ||

Ripk2
Genomic Map