siRNA Details
| Gene Symbol: Ptprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011211.3 | NP_035341.2 | 35 | 4659 | ||
| GenBank mRNA | AF326559.1 | - | 17 | 3212 | ||
| AF326560.1 | - | 17 | 3212 | |||
| BC025145.1 | - | 11 | 1504 | |||
| BC086654.1 | - | 26 | 3892 | |||
| BC141409.1 | - | 25 | 3838 | 9056022 | ||
| D13903.1 | - | 17 | 3609 | |||

Ptprd
Genomic Map