siRNA Details
| Gene Symbol: Ptprk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008983.2 | NP_033009.1 | 22 | 3205 | ||
| GenBank mRNA | BC138376.1 | - | 21 | 3135 | 8861396 | |
| BC145261.1 | - | 24 | 3213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprk| Gene Symbol: Ptprk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008983.2 | NP_033009.1 | 22 | 3205 | ||
| GenBank mRNA | BC138376.1 | - | 21 | 3135 | 8861396 | |
| BC145261.1 | - | 24 | 3213 | |||