siRNA Details
| Gene Symbol: Ptprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008984.2 | NP_033010.2 | 12 | 2450 | ||
| GenBank mRNA | AK039261.1 | - | 3 | 583 | ||
| AK129005.1 | - | 10 | 1817 | |||
| AK220171.1 | - | 10 | 1721 | |||
| BC058614.1 | - | 12 | 2460 | 30056619 | ||
| BC079621.1 | - | 12 | 2064 | 30536348 | ||
| X58287.1 | - | 12 | 2375 | |||

Ptprm
Genomic Map