siRNA Details
| Gene Symbol: Ptprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008984.2 | NP_033010.2 | 3 | 721 | ||
| GenBank mRNA | AK129005.1 | - | 1 | 88 | ||
| BC058614.1 | - | 3 | 731 | 30056619 | ||
| BC079621.1 | - | 3 | 335 | 30536348 | ||
| X58287.1 | - | 3 | 646 | |||

Ptprm
Genomic Map