siRNA Details
| Gene Symbol: Ptprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008984.2 | NP_033010.2 | 6 | 1203 | ||
| GenBank mRNA | AK129005.1 | - | 4 | 570 | ||
| AK220171.1 | - | 4 | 474 | |||
| BC058614.1 | - | 6 | 1213 | 30056619 | ||
| BC079621.1 | - | 6 | 817 | 30536348 | ||
| X58287.1 | - | 6 | 1128 | |||

Ptprm
Genomic Map