siRNA Details
| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164401.1 | NP_001157873.1 | 10 | 2291 | ||
| NM_011216.3 | NP_035346.3 | 10 | 2291 | |||
| GenBank mRNA | AF135166.1 | - | 10 | 2256 | ||
| AF295638.1 | - | 10 | 1904 | |||
| BC052743.1 | - | 10 | 2125 | 6834312 | ||

Ptpro
Genomic Map