siRNA Details
| Gene Symbol: Ptprr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011217.2 | NP_035347.1 | 2 | 659 | ||
| GenBank mRNA | AK028054.1 | - | 2 | 660 | ||
| AK051719.1 | - | 2 | 736 | |||
| BC119230.1 | - | 2 | 592 | 8733979 | ||
| BC120547.1 | - | 2 | 592 | 40129470 | ||
| D31898.1 | - | 2 | 622 | |||

Ptprr
Genomic Map