siRNA Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008991.2 | NP_033017.2 | 15 | 1389 | ||
| GenBank mRNA | AK004660.1 | - | 15 | 1370 | ||
| AK144060.1 | - | 15 | 1371 | |||
| AK153298.1 | - | 15 | 1347 | |||
| BC009119.1 | - | 15 | 1381 | 3154529 | ||
| BC054446.1 | - | 15 | 1337 | 6477711 | ||
| X89569.1 | - | 6 | 482 | |||

Abcd3
Genomic Map